(2-Chloro-6-fluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-fluorophenylacetic acid

A782647-100G A782647-1G A782647-25G A782647-5G
77244-098EA 140.08 USD
77244-098 77244-100 77244-102 77244-104
(2-Chloro-6-fluorophenyl)acetic acid 98%
(2-Chloro-6-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59 g/mol
MDL Number: MFCD00004319
CAS Number: 37777-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR