3,4-Difluorophenetole 96%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluorophenyl ethyl ether, 4-Ethoxy-1,2-difluorobenzene, 1,2-Difluoro-4-ethoxybenzene

A782545-1G A782545-25G A782545-5G
77281-296EA 8 USD
77281-296 77281-298 77281-300
3,4-Difluorophenetole 96%
3,4-Difluorophenetole

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.15
MDL Number: MFCD00673005
CAS Number: 163848-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR