3-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A781124-1G A781124-250MG A781124-5G
77281-290EA 56.65 USD
77281-290 77281-292 77281-294
3-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%
3-(Dimethylcarbamoyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₃
MW: 275.1500 g/mol
MDL Number: MFCD05863907
CAS Number: 832114-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR