AMZ30 98%

Supplier: Ambeed
Warning

Synonyms: (E)-2-(4-Fluorophenylsulphonyl)-3-(1-(3-nitrophenylsulphonyl)-1H-pyrrol-2-yl)-acrylonitrile, (E)-2-((4-Fluorophenyl)sulphonyl)-3-(1-((3-nitrophenyl)sulphonyl)-1H-pyrrol-2-yl)acrylonitrile, NIH Probe ML136

A781030-10MG A781030-1MG A781030-5MG
77244-082EA 116.39 USD
77244-082 77244-084 77244-086
AMZ30 98%
AMZ30

Precaución: for Research Use Only

Formula: C₁₉H₁₃FN₃O₆S₂
MW: 461.44 g/mol
MDL Number: MFCD28900741
CAS Number: 1313613-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR