4-Amino-1,3,5-trimethyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A780921-10G A780921-1G A780921-250MG A780921-5G
77293-682EA 187 USD
77293-682 77293-684 77293-686 77293-688
4-Amino-1,3,5-trimethyl-1H-pyrazole 98%
4-Amino-1,3,5-trimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₁₁N₃
MDL Number: MFCD00052883
CAS Number: 28466-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR