2,4,6-Trifluoroaniline 98%

Supplier: Ambeed
Danger

A780906-10G A780906-1G A780906-5G A780906-25G
77293-676EA 26.78 USD
77293-676 77293-678 77293-680 77552-424
2,4,6-Trifluoroaniline 98%
2,4,6-Trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄F₃N
MW: 147.1 g/mol
MDL Number: MFCD00007650
CAS Number: 363-81-5
UN: 2921
ADR: 8,4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR