2,2-Difluoropropionic acid ethyl ester 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2,2-Difluoropropionate, 2,2-Difluoropropionic acid ethyl ester

A780660-25G A780660-5G
77255-800EA 63 USD
77255-800 77747-528
2,2-Difluoropropionic acid ethyl ester 98%
2,2-Difluoropropionic acid ethyl ester

Precaución: for Research Use Only

Formula: C₅H₈F₂O₂
MDL Number: MFCD06657970
CAS Number: 28781-85-3
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR