4-(Bromoacetyl)benzoic acid 95%

Supplier: Ambeed
Danger

Synonyms: 4-(2-Bromoacetyl)benzoic acid

A780078-10G A780078-1G A780078-250MG A780078-25G A780078-5G
77268-084EA 128.75 USD
77268-084 77268-086 77268-088 77268-090 77268-092
4-(Bromoacetyl)benzoic acid 95%
4-(Bromoacetyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.054 g/mol
MDL Number: MFCD00229872
CAS Number: 20099-90-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR