4-Phenylbenzhydrazide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Biphenylcarboxylic acid hydrazide

A779881-100G A779881-1G A779881-25G A779881-5G
77293-666EA 381.1 USD
77293-666 77293-668 77293-670 77293-672
4-Phenylbenzhydrazide 98%
4-Phenylbenzhydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O
MW: 212.2472 g/mol
MDL Number: MFCD00017078
CAS Number: 18622-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR