H-Asn(Trt)-OtBu 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 2-amino-4-oxo-4-(tritylamino)butanoate

A779797-250MG A779797-5G A779797-1G
77244-074EA 15.45 USD
77244-074 77244-076 77747-525
H-Asn(Trt)-OtBu 97%
H-Asn(Trt)-OtBu

Precaución: for Research Use Only

Formula: C₂₇H₃₀N₂O₃
MW: 430.53 g/mol
MDL Number: MFCD30475843
CAS Number: 1998700-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR