2-Hydrazinylacetic acid 95%

Supplier: Ambeed
Danger

Synonyms: Hydrazinylacetic acid

A779046-100MG A779046-250MG
77244-070EA 38.11 USD
77244-070 77244-072
2-Hydrazinylacetic acid 95%
2-Hydrazinylacetic acid

Precaución: for Research Use Only

Formula: C₂H₆N₂O₂
MW: 90.08 g/mol
MDL Number: MFCD11168310
CAS Number: 14150-64-2
UN: 2928
ADR: 6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR