Ethyl-4-(1-piperazinyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-(piperazin-1-yl)benzoate

A778700-1G A778700-25G A778700-5G A778700-100G
77293-660EA 6 USD
77293-660 77293-662 77293-664 77749-535
Ethyl-4-(1-piperazinyl)benzoate 98%
Ethyl-4-(1-piperazinyl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD04973340
CAS Number: 80518-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR