4,4'-Butylidenebis(6-tert-butyl-m-cresol) 98%

Supplier: Ambeed
Warning

A778589-1000G A778589-100G A778589-25G A778589-500G
77293-652EA 166 USD
77293-652 77293-654 77293-656 77293-658
4,4'-Butylidenebis(6-tert-butyl-m-cresol) 98%
4,4'-Butylidenebis(6-tert-butyl-m-cresol)

Precaución: for Research Use Only

Formula: C₂₆H₃₈O₂
MDL Number: MFCD00026289
CAS Number: 85-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR