6-(4-Carboxyphenyl)nicotinic acid 97%

Supplier: Ambeed
Warning

A777402-1G A777402-250MG
77244-064EA 95 USD
77244-064 77244-066
6-(4-Carboxyphenyl)nicotinic acid 97%
6-(4-Carboxyphenyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₃H₉NO₄
MW: 243.21 g/mol
MDL Number: MFCD18317341
CAS Number: 676339-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR