FPH2 (BRD-9424) 98%

Supplier: Ambeed
Warning

Synonyms: (Z)-N-(3-carbamoyl-1-ethyl-1H-pyrazol-4-yl)-N'-(5-chloro-2-methoxyphenyl)carbamimidothioic acid, FPH2, 4-(3-(5-Chloro-2-methoxyphenyl)thioureido)-1-ethyl-1H-pyrazole-3-carboxamide

A777306-10MG A777306-1MG A777306-25MG A777306-5MG
77268-050EA 46 USD
77268-050 77268-052 77268-054 77268-056
FPH2 (BRD-9424) 98%
FPH2 (BRD-9424)

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClN₅O₂S
MDL Number: MFCD04137604
CAS Number: 957485-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR