2-(Methylthio)thiazole 98%

Supplier: Ambeed
Warning

A777198-100G A777198-5G A777198-25G
77255-784EA 96 USD
77255-784 77255-786 77618-132
2-(Methylthio)thiazole 98%
2-(Methylthio)thiazole

Precaución: for Research Use Only

Formula: C₄H₅NS₂
MDL Number: MFCD00858981
CAS Number: 5053-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR