Trimethoxy(pentafluorophenyl)silane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Trimethoxy(perfluorophenyl)silane
A777169-1G
A777169-250MG
A777169-5G
77255-778EA
42
USD
77255-778
77255-780
77255-782
Trimethoxy(pentafluorophenyl)silane 98%
Trimethoxy(pentafluorophenyl)silane
Precaución: for Research Use Only
Formula:
C₉H₉F₅O₃Si |
MDL Number:
MFCD04039205 CAS Number: 223668-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...