1-Ethoxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-naphthyl ether, 1-Ehoxynaphthalene

A776855-100G A776855-25G
77244-060EA 21.63 USD
77244-060 77244-062
1-Ethoxynaphthalene 98%
1-Ethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MW: 172.23 g/mol
MDL Number: MFCD00003928
CAS Number: 5328-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR