Trimethyl[2,3-(dioleyloxy)propyl]ammonium chloride 95%

Supplier: Ambeed
Warning

Synonyms: DOTMA, N-[1-(2,3-Dioleyloxy)propyl]-N,N,N-trimethylammonium Chloride, N,N,N-Trimethyl-2,3-bis((Z)-octadec-9-en-1-yloxy)propan-1-aminium chloride

A776776-100MG A776776-250MG A776776-50MG
77244-058EA 121.54 USD
77244-058 77618-128 77618-130
Trimethyl[2,3-(dioleyloxy)propyl]ammonium chloride 95%
Trimethyl[2,3-(dioleyloxy)propyl]ammonium chloride

Precaución: for Research Use Only

Formula: C₄₂H₈₄ClNO₂
MW: 670.59 g/mol
MDL Number: MFCD00145456
CAS Number: 104162-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR