2-(3-Biphenylyl)amino-9,9-dimethylfluorene 98%

Supplier: Ambeed
Warning

Synonyms: N-([1,1'-Biphenyl]-3-yl)-9,9-dimethyl-9H-fluoren-2-amine

A776725-1G A776725-25G A776725-5G
77244-052EA 5 USD
77244-052 77244-054 77244-056
2-(3-Biphenylyl)amino-9,9-dimethylfluorene 98%
2-(3-Biphenylyl)amino-9,9-dimethylfluorene

Precaución: for Research Use Only

Formula: C₂₇H₂₃N
MW: 361.49 g/mol
MDL Number: MFCD30471917
CAS Number: 1372778-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR