95%

Supplier: Ambeed
Warning

Synonyms: 1-Phenylhexan-3-one

A776073-100G A776073-10G A776073-25G A776073-5G
77244-044EA 72 USD
77244-044 77244-046 77244-048 77244-050
95%
1-Phenyl-3-hexanone

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂CH₂C(O)CH₂CH₂CH₃
MDL Number: MFCD00027144
CAS Number: 29898-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR