4,4'-Methylenebis(2,6-diethylaniline) 98%

Supplier: Ambeed
Warning

Synonyms: Bis(4-amino-3,5-diethylphenyl)methane, 4,4-Methylenebis(2,6-diethylaniline), 4,4'-Diamino-3,3',5,5'-tetraethyldiphenylmethane

A775959-100G A775959-25G
77293-648EA 21.63 USD
77293-648 77293-650
4,4'-Methylenebis(2,6-diethylaniline) 98%
4,4'-Methylenebis(2,6-diethylaniline)

Precaución: for Research Use Only

Formula: C₂₁H₃₀N₂
MW: 310.4763 g/mol
MDL Number: MFCD00071552
CAS Number: 13680-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR