5-Fluoro-3-methylindole 95%

Supplier: Ambeed
Warning

A775658-250MG A775658-5G A775658-1G
77281-272EA 30.9 USD
77281-272 77281-274 77573-488
5-Fluoro-3-methylindole 95%
5-Fluoro-3-methylindole

Precaución: for Research Use Only

Formula: C₉H₈FN
MW: 149.17 g/mol
MDL Number: MFCD03095377
CAS Number: 392-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR