3-(3-Thienyl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Thiophen-3-yl)propanoic acid

A774811-1G A774811-250MG A774811-5G
77293-642EA 62.83 USD
77293-642 77293-644 77293-646
3-(3-Thienyl)propanoic acid 95%
3-(3-Thienyl)propanoic acid

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.202 g/mol
MDL Number: MFCD06655385
CAS Number: 16378-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR