(2,2'-Bipyridine)bis(2-phenylpyridinato)iridium(III) Hexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: (2,2'-Bipyridine)bis[2-pyridinyl-kN)phenyl-kC]iridium(III) hexafluorophosphate

A774440-100MG A774440-250MG
77255-774EA 77.25 USD
77255-774 77255-776
(2,2'-Bipyridine)bis(2-phenylpyridinato)iridium(III) Hexafluorophosphate 98%
(2,2'-Bipyridine)bis(2-phenylpyridinato)iridium(III) Hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₂H₂₄F₆IrN₄P
MW: 801.75 g/mol
MDL Number: MFCD28976160
CAS Number: 106294-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR