4-(2-Propynyloxy)benzenecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Prop-2-yn-1-yloxy)benzaldehyde, 4-(2-Propynyloxy)benzaldehyde, 4-(Propargyloxy)benzaldehyde

A774400-10G A774400-1G A774400-25G A774400-5G A774400-100G
77293-634EA 79.31 USD
77293-634 77293-636 77293-638 77293-640 77749-531
4-(2-Propynyloxy)benzenecarbaldehyde 98%
4-(2-Propynyloxy)benzenecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD01088150
CAS Number: 5651-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR