1-(4-(3-(Piperidin-1-yl)propoxy)benzyl)piperidine 99+%

Supplier: Ambeed
Warning

Synonyms: JNJ-5207852

A774214-10MG A774214-1MG A774214-25MG A774214-50MG A774214-5MG
77268-040EA 56.65 USD
77268-040 77268-042 77268-044 77268-046 77268-048
1-(4-(3-(Piperidin-1-yl)propoxy)benzyl)piperidine 99+%
1-(4-(3-(Piperidin-1-yl)propoxy)benzyl)piperidine

Precaución: for Research Use Only

Formula: C₂₀H₃₂N₂O
MW: 316.48 g/mol
MDL Number: MFCD05974930
CAS Number: 398473-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR