1-Bromo-3-(tert-butyldimethylsiloxy)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-Bromophenoxy)(tert-butyl)dimethylsilane
A773991-1G
A773991-25G
A773991-5G
77244-038EA
16.48
USD
77244-038
77244-040
77244-042
1-Bromo-3-(tert-butyldimethylsiloxy)benzene 98%
1-Bromo-3-(tert-butyldimethylsiloxy)benzene
Precaución: for Research Use Only
Formula:
C₁₂H₁₉BrOSi MW: 287.27 g/mol |
MDL Number:
MFCD01318106 CAS Number: 65423-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...