TAS-120 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(3-(4-Amino-3-((3,5-dimethoxyphenyl)ethynyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)pyrrolidin-1-yl)prop-2-en-1-one, Futibatinib, 1-[(3S)-3-[4-amino-3-[2-(3,5-dimethoxyphenyl)ethynyl]-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-pyrrolidinyl]-2-propen-1-one

A773884-10MG A773884-1MG A773884-5MG
77268-028EA 80.34 USD
77268-028 77268-030 77268-032
TAS-120 98%
TAS-120

Precaución: for Research Use Only

Formula: C₂₂H₂₂N₆O₃
MW: 418.5 g/mol
MDL Number: MFCD29037352
CAS Number: 1448169-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR