3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-iodo-1H-pyrrolo[2,3-b]pyridine-5-carboxylate

A773434-100MG A773434-1G A773434-250MG
77281-264EA 29 USD
77281-264 77281-266 77281-268
3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid methyl ester 97%
3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₇IN₂O₂
MW: 302.07 g/mol
MDL Number: MFCD09260187
CAS Number: 944937-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR