2,4-Difluorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A772962-100G A772962-1G A772962-250MG A772962-25G A772962-5G
77244-028EA 173.04 USD
77244-028 77244-030 77244-032 77244-034 77244-036
2,4-Difluorophenylboronic acid pinacol ester 98%
2,4-Difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₂O₂
MW: 240.06 g/mol
MDL Number: MFCD05663880
CAS Number: 288101-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR