2,4-Dichloro-8-nitroquinazoline 97%

Supplier: Ambeed
Warning

A772942-100MG A772942-250MG
77244-024EA 125.66 USD
77244-024 77244-026
2,4-Dichloro-8-nitroquinazoline 97%
2,4-Dichloro-8-nitroquinazoline

Precaución: for Research Use Only

Formula: C₈H₃Cl₂N₃O₂
MW: 244.03 g/mol
MDL Number: MFCD22561791
CAS Number: 174566-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR