2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A772151-100MG A772151-1G A772151-250MG
77268-022EA 44 USD
77268-022 77268-024 77268-026
2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan 97%
2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₃₆H₂₆O₃P₂
MDL Number: MFCD30607231
CAS Number: 911397-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR