5,6-Dimethylpyrazin-2(1H)-one 97%

Supplier: Ambeed
Warning

A772021-100MG A772021-1G A772021-250MG A772021-5G
77268-014EA 15.45 USD
77268-014 77268-016 77268-018 77268-020
5,6-Dimethylpyrazin-2(1H)-one 97%
5,6-Dimethylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD13705420
CAS Number: 57229-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR