3,5-Bis(trifluoromethyl)benzamidoxime 97%

Supplier: Ambeed
Warning

Synonyms: N'-Hydroxy-3,5-bis(trifluoromethyl)benzimidamide

A771730-1G A771730-25G A771730-5G
77268-008EA 12.36 USD
77268-008 77268-010 77268-012
3,5-Bis(trifluoromethyl)benzamidoxime 97%
3,5-Bis(trifluoromethyl)benzamidoxime

Precaución: for Research Use Only

Formula: C₉H₆F₆N₂O
MW: 272.15 g/mol
MDL Number: MFCD00068188
CAS Number: 72111-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR