SGI-7079 ≥98%

Supplier: Ambeed
Warning

A771421-1MG A771421-2MG
77244-020EA 47 USD
77244-020 77244-022
SGI-7079 ≥98%
SGI-7079

Precaución: for Research Use Only

Formula: C₂₆H₂₆FN₇
MDL Number: MFCD28963942
CAS Number: 1239875-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR