1-(2-(Trifluoromethyl)pyrimidin-5-yl)ethanone 97%

Supplier: Ambeed
Warning

A771143-100MG A771143-250MG
77267-998EA 29.87 USD
77267-998 77268-000
1-(2-(Trifluoromethyl)pyrimidin-5-yl)ethanone 97%
1-(2-(Trifluoromethyl)pyrimidin-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O
MW: 190.12 g/mol
MDL Number: MFCD22062627
CAS Number: 1367970-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR