(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-(trifluoromethyl)phenylacetic acid

A771043-1G A771043-25G A771043-5G
77267-992EA 20.6 USD
77267-992 77267-994 77267-996
(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid 98%
(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00236303
CAS Number: 238754-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR