Tetrakis(4-bromophenyl)silane 97%

Supplier: Ambeed
Warning

A770831-100MG A770831-250MG A770831-5G
77255-762EA 37.08 USD
77255-762 77255-764 77255-766
Tetrakis(4-bromophenyl)silane 97%
Tetrakis(4-bromophenyl)silane

Precaución: for Research Use Only

Formula: C₂₄H₁₆Br₄Si
MW: 652.08 g/mol
MDL Number: MFCD30543388
CAS Number: 18733-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR