4,4'-Bipyridine -1,1'-dioxide 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bipyridine N,N'-dioxide, [4,4'-Bipyridine] 1,1'-dioxide

A770288-1G A770288-250MG A770288-5G
77293-626EA 36.05 USD
77293-626 77293-628 77293-630
4,4'-Bipyridine -1,1'-dioxide 98%
4,4'-Bipyridine -1,1'-dioxide

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00134383
CAS Number: 24573-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR