Bis(4-aminophenyl)sulfide 98%

Supplier: Ambeed
Danger

Synonyms: 4,4-Thiodianiline, 4,4'-Thiodianiline, 4-Aminophenyl Sulphide

A769714-10G A769714-1G A769714-25G A769714-5G
77293-618EA 39 USD
77293-618 77293-620 77293-622 77293-624
Bis(4-aminophenyl)sulfide 98%
Bis(4-aminophenyl)sulfide

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂S
MW: 216.30 g/mol
MDL Number: MFCD00025342
CAS Number: 139-65-1
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR