N-(2,6-dichloro-2'-(trifluoromethoxy)-[1,1'-biphenyl]-4-yl)-2-(4-(ethylsulfonyl)phenyl)acetamide ≥98%

Supplier: Ambeed
Warning

Synonyms: GSK805

A769555-10MG A769555-1MG A769555-25MG A769555-5MG
77244-004EA 103 USD
77244-004 77244-006 77244-008 77244-010
N-(2,6-dichloro-2'-(trifluoromethoxy)-[1,1'-biphenyl]-4-yl)-2-(4-(ethylsulfonyl)phenyl)acetamide ≥98%
N-(2,6-dichloro-2'-(trifluoromethoxy)-[1,1'-biphenyl]-4-yl)-2-(4-(ethylsulfonyl)phenyl)acetamide

Precaución: for Research Use Only

Formula: C₂₃H₁₈Cl₂F₃NO₄S
MW: 532.36 g/mol
MDL Number: MFCD28902182
CAS Number: 1426802-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR