2-(2-Chloro-4-fluorophenyl)ethanol 95%

Supplier: Ambeed
Warning

A769132-1G A769132-250MG
77244-000EA 66.95 USD
77244-000 77244-002
2-(2-Chloro-4-fluorophenyl)ethanol 95%
2-(2-Chloro-4-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.59 g/mol
MDL Number: MFCD00800610
CAS Number: 214262-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR