3,5-Dichloro-2-fluoroaniline 95%
Supplier: Ambeed
|
Warning
|
A768265-100MG
A768265-1G
A768265-250MG
A768265-5G
77255-722EA
24
USD
77255-722
77255-724
77255-726
77255-728
3,5-Dichloro-2-fluoroaniline 95%
3,5-Dichloro-2-fluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₄Cl₂FN |
MDL Number:
MFCD22495122 CAS Number: 1394838-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...