ω-Bromo-2-fluoroacetophenone 98%

Supplier: Ambeed
Danger

A768229-10G A768229-1G A768229-250MG A768229-25G A768229-5G
77243-986EA 35 USD
77243-986 77243-988 77243-990 77243-992 77243-994
ω-Bromo-2-fluoroacetophenone 98%
ω-Bromo-2-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD00278796
CAS Number: 655-15-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR