N,N'-(1R,2R)-1,2-Cyclohexanediylbis[N'-[3,5-bis(trifluoromethyl)phenyl]thiourea 98%

Supplier: Ambeed
Warning

A767953-1G A767953-250MG A767953-5G
77255-716EA 53 USD
77255-716 77255-718 77747-521
N,N'-(1R,2R)-1,2-Cyclohexanediylbis[N'-[3,5-bis(trifluoromethyl)phenyl]thiourea 98%
N,N'-(1R,2R)-1,2-Cyclohexanediylbis[N'-[3,5-bis(trifluoromethyl)phenyl]thiourea

Precaución: for Research Use Only

Formula: C₂₄H₂₀F₁₂N₄S₂
MDL Number: MFCD31707558
CAS Number: 743458-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR