Tris(3,3',3''-phosphinidynetris(benzenesulfonato)palladium(0) nonasodium salt nonahydrate 98%

Supplier: Ambeed
Warning

A767636-100MG A767636-250MG
77255-712EA 144 USD
77255-712 77255-714
Tris(3,3',3''-phosphinidynetris(benzenesulfonato)palladium(0) nonasodium salt nonahydrate 98%
Tris(3,3',3''-phosphinidynetris(benzenesulfonato)palladium(0) nonasodium salt nonahydrate

Precaución: for Research Use Only

Formula: C₅₄H₅₄Na₉O₃₆P₃PdS₉
MW: 1973.81 g/mol
MDL Number: MFCD08457632
CAS Number: 176483-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR