2,4,6-Trichloroanisole 98%

Supplier: Ambeed
Warning

A767422-25G A767422-5G
77255-708EA 38 USD
77255-708 77255-710
2,4,6-Trichloroanisole 98%
2,4,6-Trichloroanisole

Precaución: for Research Use Only

Formula: C₇H₅Cl₃O
MDL Number: MFCD00000588
CAS Number: 87-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR