4-[Methoxy(methyl)carbamoyl]benzeneboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(N,O-Dimethylhydroxylaminocarbonyl)phenylboronic acid, 4-Borono-(N-methoxy-N-methyl)benzamide
A767085-1G
A767085-25G
A767085-5G
77293-612EA
19.57
USD
77293-612
77293-614
77293-616
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid 98%
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₂BNO₄ MW: 209.007 g/mol |
MDL Number:
MFCD04115671 CAS Number: 179055-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...