6-(4-Amino-4-methylpiperidin-1-yl)-3-(2,3-dichlorophenyl)pyrazin-2-amine 98%

Supplier: Ambeed
Warning

A766981-10MG A766981-1MG A766981-25MG A766981-5MG A766981-1G A766981-100MG A766981-50MG
77243-974EA 48 USD
77243-974 77243-976 77243-978 77243-980 77747-514 77747-513 77747-515
6-(4-Amino-4-methylpiperidin-1-yl)-3-(2,3-dichlorophenyl)pyrazin-2-amine 98%
6-(4-Amino-4-methylpiperidin-1-yl)-3-(2,3-dichlorophenyl)pyrazin-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₉Cl₂N₅
MDL Number: MFCD29059453
CAS Number: 1801747-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR